Moldex™ Type N95 Particulate Respirators
Shell resists collapsing, even in heat and humidity so the respirator holds its shape longer
Manufacturer: Moldex™ 2201N95
 View more versions of this product
Catalog No. 19-003-246A
  Check Availability
Add to Cart
    Specifications
    Certifications ANSI ISEA 110-2003 7.11.1., NIOSH TC-84A-2456
    Disposable Yes
    For Use With (Application) Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
    Material Dura Mesh™
    Resistant To Non-oil based particles
    Size Small
    Filter Efficiency 95% or greater against particulate aerosols free of oil
    Color White
    Quantity 20 Pack
    Filter Type N95 Particulate Filter
    Product Type Particulate Respirator
  • Description & Specifications
Related Products
Product Content Feedback