Learn More
Moldex™ N95 Dura-Mesh™ Molded Particulate Respirators
SDP
Catalog No. 19003246E Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Color:
Blue
Gray
White
Size:
Low profile
Medium/Large
Small
With Exhalation Valve:
No
Yes
Nuisance Odor Relief:
Acid Gases
None
Ozone and Organic Vapors
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size With Exhalation Valve Nuisance Odor Relief
19-003-246E Blue Low profile Yes None
19-003-246D Blue Small Yes None
19-003-246C Blue Medium/Large Yes None
19-003-246A White Small No None
18-999-1449 White Medium/Large No None
19-003-246G Blue Medium/Large No Acid Gases
19-003-246F Gray Medium/Large No Ozone and Organic Vapors
19-003-246B White Low profile No None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-003-246E Supplier Moldex™ Supplier No. 2307N95
Add to Cart
Add to Cart
N95 respirators with latex two-strap design are constructed of a high-resistant shell that holds its shape longer even in hot and humid environments.
  • Dura-Mesh™ shell resists collapsing in heat and humidity
  • Molded nose bridge seals easily without a metal noseband
  • Facepiece is naturally contoured for less pressure
  • Straps are securely attached so they don’t break at the seal
  • Softspun™ lining for increased comfort and durability
  • Soft foam nose cushion for added comfort
  • Available in three sizes to fit more workers and increase OSHA compliance
  • NIOSH certified to have a filter efficiency of 95% or greater for non-oil based particulates
  • Certified under 42CFR84
  • Meets heat and flame resistance in accordance with ANSI/ISEA 110-2009 Section 7.11.1
  • N95 Series Applications: Grinding, torch cutting, sanding, metals, sweeping, bagging, foundries, stone quarrying, agriculture, construction, cement, ceramics manufacturing, underground mining, timber/lumber mills, polishing, buffing, lime, poultry, textiles, welding, brazing, soldering
  • N95 Plus Nuisance Levels of Ozone and Organic Vapors Applications (Fisher cat #19-003-246F only): Welding, arc cutting, foundry operations, brazing, plasma spraying, laser beam cutting/welding, laboratory settings, agriculture, petrochemical manufacturing, rendering plants, auto repair, mining operations, waste disposal
  • N95 Plus Nuisance Levels of Acid Gas Irritants Applications (Fisher cat #19-003-246G only): Aluminum smelting, glass etching, electroplating, fiberglass manufacturing, paper processing, chemical processing, brewing operations, fertilizer production
Compliance
Certified under NIOSH 42 CRF 84 to have a filter efficiency level of at least 95% effective against particulate aerosols free of oil, up to 10X the OSHA Permissable Exposure Limit (PEL).
Specifications
Aerosol Type N - Not Resistant to Oil
ANSI Standard ANSI/ISEA 110-2009, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Color Blue
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Dura-Mesh
Product Type General Purpose Particulate Respirator
Size Low profile
With Exhalation Valve Yes
Filter Efficiency 95%
For Use With (Application) Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
Latex Free No
Material Headstrap Latex
Model 2307N95
Nuisance Odor Relief None
Quantity 10/Pk.
Strap Attachment Type Two-Strap Design
FDA Cleared Surgical Mask No