Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASP MoleBio Filtered, Sterile Universal Pipette Tips

Catalog No. 21200500
Click to view available options
Volume (Metric):
10 μL
100 μL
1250 μL
20 μL
200 μL
300 μL

Universal fit tips developed to offer a reliable liquid transfer solution for molecular laboratories.

  • All theMoleBio™ Pipette tips are molded from high-quality medical grade polypropylene (PP) material, manufactured, and packed in a clean room referring to the ISO Class 5Standard, free of DNA, DNase, RNase, and PCR Inhibitor

Specifications

Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Hinged Rack Sterile
Tip Style Extended Length, Barrier, Ultra Low Retention
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.