Learn More
IBI Scientific™ Molecular Biology Grade Agarose
Molecular biology certified for use in standard electrophoresis procedures including nucleic acid, analytical, and preparative electrophoresis and blotting. IBI Scientific™ Molecular Biology Grade Agarose is highly refined and purified with strict testing parameters to insure lot-to-lot consistency.
$80.90 - $1397.00
Specifications
Product Type Specs | Agarose |
---|---|
For Use With (Application) | Electrophoresis |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
IB70041
|
IBI Scientific™
IB70041 |
250 g |
Each for $457.00
|
|
IB70035
|
IBI Scientific™
IB70035 |
25 g |
Each for $80.90
|
|
IB70045
|
IBI Scientific™
IB70045 |
1 kg |
Each for $1,397.00
|
|
IB70042
|
IBI Scientific™
IB70042 |
500 g |
Each for $813.00
|
|
IB70040
|
IBI Scientific™
IB70040 |
100 g |
Each for $243.00
|
|
Description
- Mechanical resistance
- Low absorption of staining reagents
- Transparency for high visibility
- Thermal stability due to high hysteresis
- Lot-to-lot consistency
- DNase- and RNase-Free
Specifications
Agarose | |
Electrophoresis |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.