Thermo Scientific™ Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Manufacturer: Thermo Scientific™ 3444B
 View more versions of this product
Catalog No. 21-403-182
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Microcentrifuge Tube
    Capacity (Metric) 1.5mL
    Color Blue
    Material Polypropylene
    Sterile No
    Cap Style Flat
    Graduated No
    Includes Flat cap
    Packaging Bag
    Certifications/Compliance RNase, DNase, RNA, DNA, ATP, pyrogen-free
    Closure Type Flat
  • Description & Specifications
Related Products
Product Content Feedback