Thermo Scientific™ Molecular BioProducts Snap-Cap Plain Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Thermo Scientific™ 3439G
 View more versions of this product
Catalog No. 21-403-169
  Check Availability
Add to Cart
    Specifications
    Product Type Microcentrifuge Tube
    Capacity (Metric) 0.6mL
    Color Green
    Material Polypropylene
    Sterile Yes
    Cap Style Flat
    Graduated No
    Includes Flat cap
    Packaging Bag
    Certifications/Compliance RNase, DNase, RNA, DNA, ATP, pyrogen-free
    Closure Type Flat
  • Description & Specifications
Related Products
Product Content Feedback