Learn More
Thermo Scientific™ SoftFit-L™ Non-Filtered Low Retention Pipette Tip in Reload Tower
Pipette samples with the accuracy and precision you expect from your pipette every time, with these low retention pipette tips that deliver maximum volume of sample.
$199.00 - $263.00
Specifications
Filtered | Non-Filtered |
---|---|
Color | Natural |
Clean Claims | DNase, RNase, DNA, ATP, and Endotoxin Free |
Format | Refill |
Tip Style | Universal |
Catalog Number | Mfr. No. | Description | Volume (Metric) | Color Code | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Volume (Metric) | Color Code | Price | Quantity & Availability | |||
21-402-559
|
Thermo Scientific™
372205RI |
20L, Non-Filtered, Non-sterile, Refill | 20 μL | Red |
Pack of 960 for $217.00
Case of 5 PK for $984.00
|
|
|||
21-402-570
|
Thermo Scientific™
375205RI |
200L, Non-Filtered, Non-sterile, Refill | 200 μL | Green |
Pack of 960 for $219.00
Case of 5 PK for $991.00
|
|
|||
21-402-580
|
Thermo Scientific™
373205RI |
300L, Non-Filtered, Non-sterile, Refill | 300 μL | Orange |
Pack of 960 for $199.00
Case of 5 PK for $918.00
|
|
|||
21-402-590
|
Thermo Scientific™
378205RI |
1000L, Non-Filtered, Non-sterile, Refill | 1000 μL | Blue |
Pack of 768 for $202.00
Case of 4 PK for $749.00
|
|
|||
21-402-601
|
Thermo Scientific™
374205RI |
1200L, Non-Filtered, Non-sterile, Refill | 1200 μL | Yellow |
Pack of 768 for $199.00
Case of 4 PK for $733.00
|
|
|||
21-402-557
|
Thermo Scientific™
372105RI |
20L, Non-Filtered, Sterile, Refill | 20 μL | Red |
Pack of 960 for $263.00
Case of 5 PK for $1,192.00
|
|
|||
21-402-578
|
Thermo Scientific™
373105RI |
300L, Non-Filtered, Sterile, Refill | 300 μL | Orange |
Pack of 960 for $255.00
Case of 5 PK for $1,132.00
|
|
|||
21-402-568
|
Thermo Scientific™
375105RI |
200L, Non-Filtered, Sterile, Refill | 200 μL | Green |
Pack of 960 for $263.00
Case of 5 PK for $1,192.00
|
|
|||
21-402-588
|
Thermo Scientific™
378105RI |
1000L, Non-Filtered, Sterile, Refill | 1000 μL | Blue |
Pack of 768 for $229.00
Case of 4 PK for $844.00
|
|
|||
Description
Pipette samples with the accuracy and precision you expect from your pipette every time, with Thermo Scientific™ SoftFit-L™ pipette tips. Low retention tips deliver maximum volume of sample and are for use with Rainin™ LTS pipettes.
- Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space
- Compatible with SoftFit-L empty racks
- Color-coded inserts allows quick identification of tip volume
Specifications
Non-Filtered | |
DNase, RNase, DNA, ATP, and Endotoxin Free | |
Universal | |
ART SoftFit L |
Natural | |
Refill | |
Rainin LTS LiteTouch |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.