Learn More
Christensen Education Molecular Model Organic Structure Set
Organic structure model set allows you to clearly illustrate the significance of chemical formulas.
Manufacturer: Christensen Education S440524
Includes: Molecular model sets include balls, as indicated in chart; pegs; springs; and detailed instructions. Replacement bond set includes 1.25 in. (3.18cm), wooden pegs (30), 2 in. (5.1cm) wooden pegs (10), and 2.13 in. (6.5cm) springs (10). Replacement ball sets include balls only.
Description
- Allow students to visualize the three-dimensional nature of a molecule
- Basic Set demonstrates organic structure
- Advanced Set demonstrates organic, inorganic, and molecular structure
Specifications
Assorted (Br, Cl, I, O, N) | |
Visualize the 3D nature of a molecule Demonstrates organic, inorganic and molecular structure | |
1.25 in. dia. balls: Blue/Nitrogen (3 holes); 1.125 in. dia. balls: Red/Oxygen (2 holes), green/Chlorine (1 hole) orange/Bromine (1 hole), Purple/Iodine or Fluorine | |
Molecular model basic set replacement balls; 32mm |