Learn More
Christensen Education Molecular Model Organic Structure Set
SCI_ED
Organic structure model set allows you to clearly illustrate the significance of chemical formulas.
Specifications
For Use With (Application) Visualize the 3D nature of a molecule Demonstrates organic, inorganic and molecular structure
View More Specs
Products 2
Catalog Number Mfr. No. Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Type Includes Price Quantity & Availability  
S65282
SCI_ED
View Documents
Christensen Education
S65282
Molecular Model Set 71 Balls, pegs, springs and detailed instructions This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S44052
SCI_ED
View Documents
Christensen Education
9078
Molecular Model Set 1.25 in. dia. balls: 10 - black/Carbon tetrahedral (4 holes), 2 -Blue/Nitrogen (3 holes); 1.125 in. dia. balls: 28 - yellow/Hydrogen (1 hole), 6 -red/Oxygen (2 holes), 4 - green/Chlorine (1 hole), 2 - orange/Bromine (1 hole), 2 - Purple/Iodine or Fluorine; Bonds: 30 - 1.25 in. wooden pegs, 10 - 2.125 in. pegs, 10 - 2 in. springs. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Allow students to visualize the three-dimensional nature of a molecule
  • Basic Set demonstrates organic structure
  • Advanced Set demonstrates organic, inorganic, and molecular structure
Specifications
Specifications
Visualize the 3D nature of a molecule Demonstrates organic, inorganic and molecular structure