Learn More
Invitrogen™ Molecular Probes™ Alexa Fluor™ Actin, from rabbit muscle, Alexa Fluor 568 Conjugate Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Tool for investigating cytoskeleton dynamics in vivo
Manufacturer:  Invitrogen™ A12374
Catalog No. A12374
Add to cart
Description
Description
The orange-fluorescent Alexa Fluor 568 actin conjugate is conjugate is prepared by labeling the amine residues of polymerized F-actin. After labeling, the conjugates are subjected to depolymerization and subsequent polymerization to ensure that the actin conjugates are able to assemble properly. The labeled actin that polymerizes is then separated from remaining monomeric actin by centrifugation, depolymerized and packaged in monomeric form. Supplied in solution.
Cell Analysis, Cell Structure, Cytoskeleton
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
200μg
Cytoskeletal Proteins (e.g. Actin)
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.