Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Molecular Probes™ Biotin Cadaverine (N-(5-Aminopentyl)Biotinamide, Trifluoroacetic Acid Salt) Non-distribution product as customer accommodation.

Useful in preparing biotinylated probes

Manufacturer:  Invitrogen™ A1594

Catalog No. A1594


Add to cart

Description

Description

  • Primary aliphatic amine of can be reversibly coupled to aldehydes and ketones to form Schiff base: which can be reduced to stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3)
  • Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247)

Cell Analysis, Cell Tracing and Tracking, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store at room temperature.
Biotin
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.