Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BODIPY™ FL C5-Ganglioside GM1 complexed to BSA

BSA-complexed probe can be directly dissolved in water

Supplier:  Invitrogen™ B34401

Encompass

Catalog No. B34401

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Colocalization of fluorescent cholera toxin B conjugates and the green-fluorescent BODIPY FL C5-ganglioside GM1 complexed to bovine serum albumin (BSA) observed by fluorescence microscopy provides a direct indication of the association of these molecules in lipid rafts. Complexing fluorescent lipids with BSA facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe.

Cell Analysis, Cell Structure, Golgi

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

1 mg
Sphingolipids
505/512 nm
BODIPY™
Safety and Handling

Safety and Handling

Recommended Storage : Store in freezer (-5 to -30°C) and protect from light.

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.