Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myc Tag Chicken anti-Tag, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen A21281

Catalog No. A21281


Add to cart

Description

Description

c-Myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors. Translocation of the c-myc locus on chromosome 8 to the immunoglobulin loci on chromosome 14 (heavy chain); 2 (delta light chain); or 22 (light chain) is described in Burkitts lymphoma and other B-cell lymphoproliferative conditions. An aberrant expression of the c-myc gene occurs in tumors of different origins such as colorectal, gastric, gallbladder, hepatic, mammary, ovarian, endometrial, head and neck, pulmonary, prostatic, thyroidal, oral, ocular, nasopharyngeal, endocrine, as well as hematopoietic neoplasms.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

Myc Tag
Polyclonal
Unconjugated
MYC
Liquid
MYC
Was raised against the peptide EQKLISEEDL.
Purified
RUO
Antibody
Polyclonal
Tag
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
P01106
c Myc, Cellular myelocytomatosis oncogene, Myc protein, Myc proto oncogene protein, Myc2, Niard, Nird, v myc avian myelocytomatosis viral oncogene hom
Chicken
IgY
100 μL
4° C
Primary
4609
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.