Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CBQCA Protein Quantitation Kit
Extremely sensitive assay for quantitating proteins in solution, capable of detection as low as 10ng of protein per mL
Supplier: Invitrogen™ C6667
Description
- Similar in sensitivity to NanoOrange protein quantitation reagent (N-6666)
- CBQCA is better suited for accurate quantitation of proteins in presence of lipids, membrane fractions or detergents, and for lipoproteins and small peptides
Protein Quantitation, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Fluorescence | |
Quantitating proteins in solution | |
Room Temperature |
Fluorescent Protein Assay | |
Protein Quantitation Assay | |
Microplate Reader | |
Store in freezer (-5 to -30°C) and protect from light. |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction