Learn More
Invitrogen™ Molecular Probes™ DMNPE-caged ATP (Adenosine 5'-Triphosphate, P3-(1-(4,5-Dimethoxy-2-Nitrophenyl)ethyl) Ester, Disodium Salt) Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Flash photolysis of the caging group yields highly localized release of the free nucleotide
Manufacturer:  Invitrogen™ A1049
Catalog No. A1049
Add to cart
Description
Description
DMNPE-caged ATP is a nucleotide analog in which the terminal phosphate is esterified with a blocking group, rendering the molecule biologically inactive. Flash photolysis of the caging group with UV illumination at 355nm results in a rapid and highly localized release of the free nucleotide at the site of illumination. The DMNPE caging group absorbs light more efficiently than CNB and NPE caging groups at approx. 360nm, but photolysis rates and quantum yields are generally lower.
Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Shipping Condition: Wet ice
Specifications
Specifications
5mg
Store in freezer (-5° to -30°C) and protect from light.
Nucleotide, Caged
DMNPE (P3-(1-(4,5-dimethoxy-2-nitrophenyl)ethyl) ester)
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.