Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

GFP Mouse anti-Tag, Clone: 11E5, Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A11121

Catalog No. A11121



Description

Description

The anti-GFP antibody is suited for detection of native GFP, GFP variants, and most GFP-fusion proteins by western blot analysis. Anti-GFP mouse monoclonal antibody 11E5 is optimized for western analysis, allowing colorimetric detection of as little as 10 ng of GFP or GFP-fusion proteins, or chemiluminescent detection of picogram quantities. These antibodies have been purified from mouse hybridoma supernatants by protein G chromatography and stabilized with BSA before lyophilization. At the time of preparation, the products are certified to be free of unconjugated dyes. They are tested in a cytological experiment to ensure low nonspecific staining. To prepare 0.2 mg/mL stock solutions, reconstitute the lyophilized antibodies in 0.5 mL of phosphate-buffered saline (PBS), pH 7.4. You may store these solutions for up to 3 months at 4°C with the addition of 2 mM sodium azide.

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. Green Fluorescent Protein (GFP), from the jellyfish Aequorea victoria, has quickly become a versatile marker for monitoring physiological processes, visualizing protein localization, and detecting transgenic expression, and a powerful research tool for assessing gene expression and subcellular protein distribution in fixed or living cells. GFP is excited by and brightly fluoresces when exposed to UV or blue light. This feature makes it ideal as a marker for use in fluorescence microscopy, cytometry, tagging fusion proteins, and assaying transcriptional regulation from gene promoters in vivo. Numerous GFP variants with enhanced and shifted emission spectra (blue, green, and yellow) have been developed through amino acid substitutions at specific residues.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

GFP
Monoclonal
Unconjugated
Lyophilized
Mouse
IgG1
100 μg
-20°C
Primary
Tag
Western Blot
11E5
Antibody with BSA and no preservative
GFP2; Green Fluoresecent Protein; GFP; Turbo GFP; eGFP; GFP tag
The GFP was isolated directly from the jellyfish Aequorea victoria.
Protein G
RUO
Antibody
Monoclonal
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.