Learn More
Invitrogen™ Glutathione Agarose, Linked Through Sulfur, sedimented bead suspension Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
For purification of GST fusion proteins
Manufacturer:  Invitrogen™ G2879
Catalog No. G2879
Add to cart
Description
Description
Glutathione agarose prepared by linking the sulfur atom of glutathione to crosslinked beaded agarose can be used for purification of GST fusion proteins. Each milliliter of gel can bind approximately 5-6mg of bovine-liver GST. Adding excess free glutathione liberates the GST fragment from the matrix which can then be regenerated by washing with a high-salt buffer.
GST-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
Affinity Column
Wet Ice
Suspension
Store in refrigerator (2–8°C).
Glutathione
SDS
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.