Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Mouse anti-Tag, Alexa Fluor® 594, Clone: 16B12, Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A21288

Catalog No. A21288


Add to cart

Description

Description

HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

HA Tag
Monoclonal
1 mg/mL
PBS with 0.1% BSA and 5mM sodium azide; pH 7.2
Human influenza hemagglutinin; hemagglutinin; HA-tag; HA; HA Epitope Tag
twelve amino acid peptide CYPYDVPDYASL.
Purified
RUO
Antibody
Monoclonal
Western Blot
16B12
Alexa Fluor™ 594
Liquid
Mouse
IgG1
100 μL
4° C, store in dark
Primary
Tag
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.