Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HPTS; pyranine (8-Hydroxypyrene-1,3,6-Trisulfonic Acid, Trisodium Salt) Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Water-soluble membrane-impermeant pH indicator

Manufacturer:  Invitrogen™ H348

Catalog No. H348


Add to cart

Description

Description

  • pKa of approximately 7.3 in aqueous buffers

Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Ionic Homeostasis and Signaling, pH Detection

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Green
Store at room temperature and protect from light.
Room Temperature
Fluorescence Microscope
Fluorescent
Cytoplasm & Cytosol
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.