Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieve, Activated, Type 4A, 8 to 12 Mesh, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle

Chemical Identifiers

1344-01-0
AlCaNaO4Si+2
182.13
IDZYHGDLWGVHQM-UHFFFAOYSA-N
44144394

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 2.10kg
Quantity 2.5kg
Packaging Wide Mouth PVC Coated Glass Bottle
Mesh Size 8 to 12 Mesh
SMILES [O-][Si]([O-])([O-])[O-].[Na+].[Al+3].[Ca+2]
IUPAC Name aluminum;calcium;sodium;silicate
Chemical Name or Material Molecular Sieve, Activated, Type 4A
Grade BAKER ANALYZED™ Reagent