Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieve Type 3A, MilliporeSigma™
GSA_VA

Click to view available options
Quantity:
500g
Packaging:
Glass Bottle
1313-59-3
Na2O
61.979
KKCBUQHMOMHUOY-UHFFFAOYSA-N
73971
disodium;oxygen(2-)
[O-2].[Na+].[Na+]
1313-59-3
Na2O
61.979
KKCBUQHMOMHUOY-UHFFFAOYSA-N
73971
disodium;oxygen(2-)
[O-2].[Na+].[Na+]

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 500g
Packaging Glass Bottle
Linear Formula Na2O, K2O, SiO2, Al2O3, MgO
Physical Form Pellet
Chemical Name or Material Molecular Sieve, Type 3A (1/16)