Alfa Aesar™ Molecular sieve, Type 4A

Manufacturer: Alfa Aesar™ 44557A7

Quantity 5kg

 View more versions of this product

Catalog No. AA44557A7

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00147627
    Chemical Name or Material Molecular sieve, Type 4A

    Specifications

    MDL Number MFCD00147627
    Mesh Size 4 to 8 Mesh
    Quantity 5kg
    TSCA TSCA
    Physical Form Beads
    Chemical Name or Material Molecular sieve, Type 4A

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit