Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Molecular Sieve Uop Type 3A, Honeywell Fluka™

Beads

$53.67 - $159.07

Specifications

Chemical Name or Material Molecular Sieve Uop Type 3A
UN Number NONH for all modes of transport
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
60-018-05 View Documents Honeywell Fluka™
02573250G
250g Each for $53.67
Add to cart
 
60-018-04 View Documents Honeywell Fluka™
025731KG
1kg Each for $159.07
Add to cart
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Molecular Sieve Uop Type 3A
NONH for all modes of transport
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit