Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 13X, 4 to 8 mesh, ACROS Organics™

Click to view available options
Quantity:
500g
5kg
Packaging:
Glass bottle
Plastic drum
63231-69-6
MFCD00131613
63231-69-6
MFCD00131613
Drying agent
Color Beige to Brown
Quantity 500g
Residual Water 1.5% max.
Bulk Density 600g/L min.
Packaging Glass bottle
Adsorption CO2-adsorption: 11.5% min.
Physical Form Beads
Chemical Name or Material Molecular sieves 13X, 13X, 4 to 8 mesh