Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Molecular sieves, 3A, 1-2mm (0.04-0.08in) beads

Click to view available options
Quantity:
250g
1000g
5000g
308080-99-1
AlNaO6Si2
202.135
MFCD00131613
URGAHOPLAPQHLN-UHFFFAOYSA-N
19758701
aluminum;sodium;dioxido(oxo)silane
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
308080-99-1
AlNaO6Si2
202.135
MFCD00131613
URGAHOPLAPQHLN-UHFFFAOYSA-N
19758701
aluminum;sodium;dioxido(oxo)silane
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Sensitivity Hygroscopic
Quantity 250g
Solubility Information Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone.
Chemical Name or Material Molecular sieves, 3A, 1 to 2mm (0.04 to 0.08 in) beads

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen