Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 3A, powder <50 micron, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
10kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
308080-99-1
AlNaO6Si2
202.135
MFCD00131613
URGAHOPLAPQHLN-UHFFFAOYSA-N
19758701
aluminum;sodium;dioxido(oxo)silane
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
308080-99-1
AlNaO6Si2
202.135
MFCD00131613
URGAHOPLAPQHLN-UHFFFAOYSA-N
19758701
aluminum;sodium;dioxido(oxo)silane
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
Drying agent
Color Undesignated
Quantity 10kg
pH 11.4 (10% max. aq. suspension)
Particle Size 500 Micron Max.
Water Adsorption > 22% (50%rH, 20°C, 24h)
Packaging Plastic drum
Additional Information 3 % max. (550°C, 2 h)
Residual Water < 1.5% (550°C, 2 hrs.)
Solubility Information Solubility in water: insoluble.
Physical Form Powder or Crystals
Chemical Name or Material Molecular sieves 3A
Show More Show Less