Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 4A, 4 to 8 mesh, ACROS Organics™

Click to view available options
Quantity:
500g
5kg
Packaging:
Glass bottle
Plastic drum
70955-01-0
C20H25FN2O8
440.424
MFCD00131613
FJUZEZRZKVMAMD-DRZCJDIDSA-N
73906282
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
CC(C(C(=O)OC)C(=O)OC)C(C(=O)O)NC(=O)C(CC1=CC(=CC=C1)F)NC(=O)C
70955-01-0
C20H25FN2O8
440.424
MFCD00131613
FJUZEZRZKVMAMD-DRZCJDIDSA-N
73906282
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
CC(C(C(=O)OC)C(=O)OC)C(C(=O)O)NC(=O)C(CC1=CC(=CC=C1)F)NC(=O)C
Drying agent
Color Colorless to Yellow
Quantity 500g
Water Adsorption 20% min.
Additional Information 1.5 % max.
Residual Water 1.5% max.
Bulk Density 660 to 760g/L
Packaging Glass bottle
Physical Form Solution
Chemical Name or Material Molecular sieves 4A