Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 4A, powder <50 micron, ACROS Organics™

Click to view available options
Quantity:
25g
500g
1kg
2.5kg
10kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
70955-01-0
C20H25FN2O8
440.424
MFCD00131613
FJUZEZRZKVMAMD-DRZCJDIDSA-N
73906282
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
CC(C(C(=O)OC)C(=O)OC)C(C(=O)O)NC(=O)C(CC1=CC(=CC=C1)F)NC(=O)C
70955-01-0
C20H25FN2O8
440.424
MFCD00131613
FJUZEZRZKVMAMD-DRZCJDIDSA-N
73906282
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
CC(C(C(=O)OC)C(=O)OC)C(C(=O)O)NC(=O)C(CC1=CC(=CC=C1)F)NC(=O)C
Drying agent
Color White to Beige
Quantity 25g
pH 9 to 12 (10% water suspension)
Residual Water 3% max.
Packaging Glass bottle
Water Adsorption 18% max.
Solubility Information Solubility in water: insoluble.
Physical Form Powder
Chemical Name or Material Molecular sieves 4A, Powder, <50 micron