Alfa Aesar™ Molecular sieves, tantalum oxide based; mesoporous, 22 Angstrom, powder

Catalysis, gas adsorption, optoelectronic materials

Manufacturer: Alfa Aesar™ 4261206

Quantity 5g

 View more versions of this product

Catalog No. AA4261206

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00131613
    Synonym Ta TMSI
    Chemical Name or Material Molecular sieves, tantalum oxide based, Mesoporous

    Specifications

    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00131613
    Synonym Ta TMSI
    Particle Size 22Å
    Sensitivity Hygroscopic
    Quantity 5g
    Chemical Name or Material Molecular sieves, tantalum oxide based, Mesoporous

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit