Alfa Aesar™ Molecular sieves, tantalum oxide based; mesoporous, 32 Angstrom, powder

Manufacturer: Alfa Aesar™ 42611-06

Quantity 5g

 View more versions of this product

Catalog No. AA4261106

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00131613
    Synonym Ta-TMSI
    Chemical Name or Material Molecular sieves, tantalum oxide based, Mesoporous

    Specifications

    Physical Form Powder
    TSCA TSCA
    MDL Number MFCD00131613
    Synonym Ta-TMSI
    Particle Size 32Å
    Quantity 5g
    Chemical Name or Material Molecular sieves, tantalum oxide based, Mesoporous

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit