Learn More
Alfa Aesar™ Molecular sieves, titanium oxide based; mesoporous, 25 Angstrom, powder
In oxidation and photocatalysis
Manufacturer:  Alfa Aesar™ 4325004
Quantity 2g
 View more versions of this product
Catalog No. AA4325004
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00131613
Ti-TMS1; Mesoporous titanium oxide based molecular sieve
Molecular sieves, titanium oxide based, Mesoporous
Specifications
25Å
2g
MFCD00131613
Molecular sieves, titanium oxide based, Mesoporous
Hygroscopic
Powder
Ti-TMS1; Mesoporous titanium oxide based molecular sieve
Safety and Handling
SDS & Safety Documents
GHS H Statement
H351-H332
Suspected of causing cancer.
Harmful if inhaled.
GHS P Statement
P261-P281-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA