Learn More
Moore Medical Molnlycke Health Care™ Hibiclens™ Liquid Antiseptic
Antimicrobial skin cleanser possessing bactericidal activities
Specifications
Quantity Each
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
19-898-789
View Documents
Moore Medical
84282
Hibiclens Liquid Antiseptic This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-898-988
View Documents
Moore Medical
92708
Hibiclens Liquid Antiseptic Pump This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description
Description
  • Non-drying
  • Low skin irritation
  • Contains 4% w/v Hibitane (chlorhexidine gluconate)
Indicated for use as a surgical scrub, a patient pre-operative skin preparation, a healthcare personnel hand wash, a skin wound and general skin cleaner.
Specifications
Specifications
Each