Molnlycke Health Care™ Hibiclens™ Liquid Antiseptic

Antimicrobial skin cleanser possessing bactericidal activities

Manufacturer: Moore Medical Corp 84282

 View more versions of this product

Catalog No. 19-898-789

  Check Availability
Add to Cart

    Specifications

    Size (English) 4 oz.
    Size (Metric) 118mL
    Quantity Each
    Composition Chlorhexidine Gluconate 4% w/v, chemically unique Hexamethylenebis Biguanide with inactive ingredients: Fragrance, Isopropyl Alcohol 4%, purified Water, red 40, other ingredients
    For Use With (Application) For use as a surgical scrub, patient pre-operative skin preparation, healthcare personnel hand wash
    Product Type Hibiclens Liquid Antiseptic
    pH Range 5 to 6.5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit