Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum boat; Thickness (mm), 0.05; Length (mm), 75, Alfa Aesar™

Click to view available options
Quantity:
1 ea.
5 ea.
Packaging Carton
Quantity 5 ea.
Physical Form Solid
Chemical Name or Material Molybdenum boat; Thickness (mm)

TSCA : U

Recommended Storage : Ambient temperatures