Alfa Aesar™ Molybdenum boride, 99% (metals basis)

Manufacturer: Alfa Aesar™ 1256314

Quantity 25g

 View more versions of this product

Catalog No. AA1256314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12006-98-3
    Formula Weight 106.75
    MDL Number MFCD00014219
    Chemical Name or Material Molybdenum boride

    Specifications

    Formula Weight 106.75
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00014219
    Quantity 25g
    CAS 12006-98-3
    Chemical Name or Material Molybdenum boride
    Assay 99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit