Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum carbide, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1219236

Quantity 500g

 View more versions of this product

Catalog No. AA1219236


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12069-89-5
99.5% (metals basis)
203.91
Molybdenum carbide
Powder
Mo2C
MFCD00014218

Specifications

500g
White powder
99.5% (metals basis)
203.91
Odorless
Molybdenum carbide
12069-89-5
Powder
Mo2C
MFCD00014218
White
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit