Molybdenum etchant, Alfa Aesar™

Manufacturer: Alfa Aesar 44768R0

Quantity 1 qt.

 View more versions of this product

Catalog No. AA44768R0

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    MDL Number MFCD08064551
    Chemical Name or Material Molybdenum etchant

    Specifications

    Physical Form Liquid
    Quantity 1 qt.
    Color Amber
    DOT Information DOT Class: 8, Packaging Group: III
    MDL Number MFCD08064551
    Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Molybdenum etchant

    H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit