Alfa Aesar™ Molybdenum gauze, 165 mesh woven from 0.0509mm (0.002 in.) dia. wire, twilled weave

Manufacturer: Alfa Aesar™ 42404HB

Quantity 300x300mm

 View more versions of this product

Catalog No. AA42404HB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-98-7
    MDL Number MFCD00003465
    Chemical Name or Material Molybdenum gauze, 165 mesh woven from 0.0509mm (0.002 in.) dia. wire, twilled weave

    Specifications

    TSCA TSCA
    Physical Form Wire Cloth
    MDL Number MFCD00003465
    CAS 7439-98-7
    Mesh Size 165 Mesh
    Quantity 300x300mm
    Chemical Name or Material Molybdenum gauze, 165 mesh woven from 0.0509mm (0.002 in.) dia. wire, twilled weave

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit