Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Molybdenum gauze, 35 mesh woven from 0.0509mm (0.002 in.) dia. wire

Manufacturer:  Alfa Aesar™ 42485GV

Quantity 150x600mm

 View more versions of this product

Catalog No. AA42485GV

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7439-98-7
MFCD00003465
Molybdenum gauze, 35 mesh woven from 0.0509mm (0.002 in.) dia. wire

Specifications

150x600mm
Wire Cloth
35 Mesh
7439-98-7
MFCD00003465
Molybdenum gauze, 35 mesh woven from 0.0509mm (0.002 in.) dia. wire

Safety and Handling

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit