Learn More
Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Description
Molybdenum(IV) oxide is used to catalyze the dehydrogenation of alcohols. It is also used as an intermediates and processing aids. It finds application in the manufacture of molybdenum nano-wires.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Red to Brown |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99% (metals basis) |
Formula Weight | 127.94 |
Percent Purity | 99% |
Odor | Odorless |
Density | 6.47 g/cm3 at 20°C |
Assay | (metals basis) |
Chemical Name or Material | Molybdenum(IV) oxide |
Safety and Handling
Hazard Category | H301-H332-H319-H335: Toxic if swallowed. Harmful if inhaled. Causes serious eye irritation. May cause respiratory irritation. |
Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of insufficient ventilation, wear suitable respiratory equipment. |
EINECSNumber | 242-637-9 |
RTECSNumber | QA4688000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.