Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum(IV) sulfide, 98.5%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic drum
1317-33-5
MoS2
160.07
CWQXQMHSOZUFJS-UHFFFAOYSA-N
molybdenum disulfide,molybdenite,molybdenum iv sulfide,molybdenum disulphide,molysulfide,molykote,motimol,nichimoly c,sumipowder pa,molykote z
14823
CHEBI:30704
bis(sulfanylidene)molybdenum
S=[Mo]=S
1317-33-5
MoS2
160.07
CWQXQMHSOZUFJS-UHFFFAOYSA-N
molybdenum disulfide,molybdenite,molybdenum iv sulfide,molybdenum disulphide,molysulfide,molykote,motimol,nichimoly c,sumipowder pa,molykote z
14823
CHEBI:30704
bis(sulfanylidene)molybdenum
S=[Mo]=S
Assay Percent Range 98% min. (MoS2)
Linear Formula MoS2
Merck Index 15,6322
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in aqua regia and sulfuric acid
Formula Weight 160.07
Physical Form Powder
Percent Purity 98.5%
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in aqua regia and sulfuric acid
Color Black or Gray
Melting Point 2375.0°C
Quantity 100g
Chemical Name or Material Molybdenum(IV) sulfide
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 215-263-9