Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Molybdenum(IV) sulfide, 98.5%, ACROS Organics™
Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic drum
Specifications
Assay Percent Range | 98% min. (MoS2) |
Linear Formula | MoS2 |
Merck Index | 15,6322 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in aqua regia and sulfuric acid |
Formula Weight | 160.07 |
Physical Form | Powder |
Percent Purity | 98.5% |
Packaging | Glass bottle |
Solubility | Solubility in water: insoluble. Other solubilities: soluble in aqua regia and sulfuric acid |
Color | Black or Gray |
Show More |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 215-263-9