Alfa Aesar™ Molybdenum powder, -250 mesh, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 0093218

Quantity 50g

 View more versions of this product

Catalog No. AA0093218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-98-7
    MDL Number MFCD00003465
    UN Number UN3089
    Chemical Name or Material Molybdenum powder

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    CAS 7439-98-7
    Mesh Size −250 Mesh
    Color Gray
    MDL Number MFCD00003465
    UN Number UN3089
    Odor Odorless
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    TSCA TSCA
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    RTECS Number QA4680000
    Quantity 50g
    Health Hazard 1 Warning
    Chemical Name or Material Molybdenum powder
    Assay 99.9%, (metals basis)

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit