Alfa Aesar™ Molybdenum silicide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 41079KS

Quantity 1 each

 View more versions of this product

Catalog No. AA41079KS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12136-78-6
    Formula Weight 152.13
    MDL Number MFCD00151535
    Chemical Name or Material Molybdenum silicide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick

    Specifications

    Formula Weight 152.13
    Density 6.31g/mL
    MDL Number MFCD00151535
    Odor Odorless
    Quantity 1 each
    Physical Form Target
    CAS 12136-78-6
    Color Dark gray
    TSCA TSCA
    Melting Point 1870° to 2030°C
    Chemical Name or Material Molybdenum silicide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
    Assay 99.5%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit