Alfa Aesar™ Molybdenum Thinfoil, 0.0025mm (0.0001 in.) thick, 99.9% (metals basis)

Microelectronics, coatings, high temperature materials

Manufacturer: Alfa Aesar™ 40710FF

Quantity 25x25mm

 View more versions of this product

Catalog No. AA40710FF

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7439-98-7
    MDL Number MFCD00003465
    Chemical Name or Material Molybdenum Thinfoil, 0.0025mm (0.0001 in.) thick, Not certified pinhole free

    Specifications

    Physical Form Foil
    MDL Number MFCD00003465
    TSCA TSCA
    CAS 7439-98-7
    Weight ≈0.016g/25x25mm
    Quantity 25x25mm
    Chemical Name or Material Molybdenum Thinfoil, 0.0025mm (0.0001 in.) thick, Not certified pinhole free
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit