Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum(V) chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
500g
Packaging:
Glass bottle
10241-05-1
Cl5Mo
273.2
MFCD00003467
GICWIDZXWJGTCI-UHFFFAOYSA-I
molybdenum v chloride,molybdenum pentachloride,molybdenum chloride mocl5,mocl5,pentachloridomolybdenum,molybdenum 5+ chloride,acmc-1bx8l,ksc491a7h
61497
CHEBI:30635
pentachloromolybdenum
Cl[Mo](Cl)(Cl)(Cl)Cl
10241-05-1
Cl5Mo
273.2
MFCD00003467
GICWIDZXWJGTCI-UHFFFAOYSA-I
molybdenum v chloride,molybdenum pentachloride,molybdenum chloride mocl5,mocl5,pentachloridomolybdenum,molybdenum 5+ chloride,acmc-1bx8l,ksc491a7h
61497
CHEBI:30635
pentachloromolybdenum
Cl[Mo](Cl)(Cl)(Cl)Cl
C-X bond formation reagent
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula MoCl5
UN Number 2508
Fieser 06,412; 01,1015; 02,365; 03,250; 04,428; 05,587; 06,519; 08,443; 09,414; 11,471; 12,434; 13,273; 04,99; 16,95
Formula Weight 273.2
Physical Form Fine Crystals or Crystalline Powder
Percent Purity ≥94%
Packaging Glass bottle
Color Black to Blue or Black-Green
Quantity 500g
Chemical Name or Material Molybdenum(V) chloride, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. Reacts violently with water. Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger

EINECSNumber : 233-575-3

RTECSNumber : QA4690000

TSCA : TSCA