Alfa Aesar™ Molybdenum(V) trichloride-isopropoxide, 25% (w/v) in isopropanol

Manufacturer: Alfa Aesar™ 44474AB

Quantity 10mL

 View more versions of this product

Catalog No. AA44474AB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 53780-81-7
    Molecular Formula MoCl3(OC3H7)2
    Formula Weight 320.48
    MDL Number MFCD02682829
    UN Number UN2924
    Chemical Name or Material Molybdenum (V) trichloride-isopropoxide

    Specifications

    Formula Weight 320.48
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    CAS 53780-81-7
    Sensitivity Moisture Sensitive
    Concentration 25% (w/v) in isopropanol
    Physical Form Liquid
    Molecular Formula MoCl3(OC3H7)2
    UN Number UN2924
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    MDL Number MFCD02682829
    Health Hazard 1 Danger
    Quantity 10mL
    Chemical Name or Material Molybdenum (V) trichloride-isopropoxide

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit