Alfa Aesar™ Molybdenum(VI) oxide, 99.5%

Manufacturer: Alfa Aesar™ A1115930

Quantity 250g

 View more versions of this product

Catalog No. AAA1115930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1313-27-5
    Molecular Formula MoO3
    Formula Weight 143.94
    MDL Number MFCD00003469
    UN Number UN3288
    Merck Index 14,6239
    Synonym Molybdenum trioxide
    Chemical Name or Material Molybdenum (VI) oxide

    Specifications

    Formula Weight 143.94
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00003469
    Merck Index 14,6239
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Odor Odorless
    CAS 1313-27-5
    Boiling Point 1155°C (subl.)
    Melting Point 795°C
    Health Hazard 2 GHS H Statement
    H300-H351-H319-H335
    Fatal if swallowed.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula MoO3
    UN Number UN3288
    Quantity 250g
    Health Hazard 1 Danger
    RTECS Number QA4725000
    TSCA TSCA
    Synonym Molybdenum trioxide
    Chemical Name or Material Molybdenum (VI) oxide
    Assay 99.5%

    Description

    Applications
    Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in the oil industry. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.

    Solubility
    Slightly soluble in water. Soluble in concentrated mineral acids and alkali hydroxide solutions.

    Notes
    It does not react with acids other than hydrofluoric and concentrated sulfuric acids.

    GHS H Statement
    H300-H351-H319-H335
    Fatal if swallowed.
    Suspected of causing cancer.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit