Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum(VI) oxide, 99.999%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1313-27-5
MoO3
143.947
JKQOBWVOAYFWKG-UHFFFAOYSA-N
molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide
14802
CHEBI:30627
trioxomolybdenum
O=[Mo](=O)=O
1313-27-5
MoO3
143.947
JKQOBWVOAYFWKG-UHFFFAOYSA-N
molybdenum trioxide,molybdenum vi oxide,molybdic anhydride,molybdic trioxide,molybdena,molybdic oxide,molybdenum oxide,molybdic acid anhydride,molybdenum peroxide
14802
CHEBI:30627
trioxomolybdenum
O=[Mo](=O)=O
For ultra trace analysis
CAS Min % 100.0
CAS Max % 100.0
Assay Percent Range 99.999%
Linear Formula MoO3
Fieser 04,203; 13,279; 16,227
Formula Weight 143.94
Physical Form Powder
Percent Purity 64.3 to 69.0%
Grade Trace Metal Basis
Packaging Glass bottle
Total Trace Metal Impurities 10ppm max.
Color Gray-Green to Green
Quantity 5g
Chemical Name or Material Molybdenum(VI) oxide
Show More Show Less

GHS H Statement:
Suspected of causing cancer.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning