Molybdenum(VI) oxide, 99.9995% (metals basis excluding W), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1293022

Quantity 100g

 View more versions of this product

Catalog No. AA1293022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1313-27-5
    Physical Form Powder
    Assay Percent Range 99.9995% (metals basis excluding W)
    Molecular Formula MoO3
    Formula Weight 143.94
    MDL Number MFCD00003469
    Chemical Name or Material Molybdenum(VI) oxide

    Specifications

    Physical Form Powder
    Formula Weight 143.94
    Quantity 100g
    Color White to gray
    DOT Information Hazard Class: 6.1, Packing Group: III
    Density 4.692g/cm3
    EINECS Number 215-204-7
    Odor Odorless
    Assay Percent Range 99.9995% (metals basis excluding W)
    Molecular Formula MoO3
    Appearance White to gray powder
    CAS 1313-27-5
    MDL Number MFCD00003469
    Health Hazard 1 H300-H319-H335-H351: Fatal if swallowed. Causes serious eye irritation. May cause respiratory irritation. Suspected of causing cancer.
    Chemical Name or Material Molybdenum(VI) oxide

    H300-H319-H335-H351: Fatal if swallowed. Causes serious eye irritation. May cause respiratory irritation. Suspected of causing cancer.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit